Learn More
Medchemexpress LLC G-5555 hydrochloride | 2319590-15-1 | 98.3% | 529.42 | 25 MG
SDP
Catalog No. 5000403040
Encompass_Preferred
G-5555 hydrochloride is a potent and selective p21-activated kinase 1 (PAK1) inhibitor with a Ki of 3.7 nM. This compound is intended for research use only.
  • Potent and selective p21-activated kinase 1 (PAK1) inhibitor.
  • Exhibits excellent kinase selectivity.
  • Shows low blood clearance and acceptable half-life.
  • Offers good oral exposure and high oral bioavailability.
  • Demonstrates tumor growth inhibition in xenograft models.
Catalog No. 50-004-03040 Supplier Medchemexpress LLC Supplier No. 25MGHY19635A
G-5555 hydrochloride is a potent and selective p21-activated kinase 1 (PAK1) inhibitor with a Ki of 3.7 nM. This compound is intended for research use only.
  • Potent and selective p21-activated kinase 1 (PAK1) inhibitor.
  • Exhibits excellent kinase selectivity.
  • Shows low blood clearance and acceptable half-life.
  • Offers good oral exposure and high oral bioavailability.
  • Demonstrates tumor growth inhibition in xenograft models.