Learn More
Electron Microscopy Sciences Pharmaceutical PSA Pattern Graticle G-57, 19 mm, Glass sandwich
SDP
Catalog No. 5027836
Encompass_Preferred
This graticule was designed for the pharmaceutical industry. However, it is also useful where particle size considerations are restricted to 10 mm and 25 mm. Dots and circles give quick references for these two sizes. In addition a scale is incorporated. The microscope must be calibrated when ordering this graticule, such that the circle must equate to 1 mm on the microscope stage.
Size: 19 mm
Surface: Glass sandwich
Catalog No. 50-278-36 Supplier Electron Microscopy Sciences Supplier No. 6809119
Add to Cart
Add to Cart
This graticule was designed for the pharmaceutical industry. However, it is also useful where particle size considerations are restricted to 10 mm and 25 mm. Dots and circles give quick references for these two sizes. In addition a scale is incorporated. The microscope must be calibrated when ordering this graticule, such that the circle must equate to 1 mm on the microscope stage.
Size: 19 mm
Surface: Glass sandwich