Learn More
Medchemexpress LLC HY-12333 5mg Medchemexpress, G-749 CAS:1457983-28-6 Purity:>98%
SDP
Catalog No. 502081012
Encompass_Preferred
Medchemexpress, HY-12333 5mg G-749 CAS:1457983-28-6 G-749 is a potent, ATP competitive FLT3 inhibitor, with IC50s of 0.4 nM and 0.6 nM for FLT3WT and FLT3D835Y, respectively. G-749 can be used for the research of drug resistance for AML (acute myeloid leukemia). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1012 Supplier Medchemexpress LLC Supplier No. HY123335MG
Medchemexpress, HY-12333 5mg G-749 CAS:1457983-28-6 G-749 is a potent, ATP competitive FLT3 inhibitor, with IC50s of 0.4 nM and 0.6 nM for FLT3WT and FLT3D835Y, respectively. G-749 can be used for the research of drug resistance for AML (acute myeloid leukemia). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.