Learn More
Medchemexpress LLC HY-N0067 100mg Medchemexpress, γ-Aminobutyric acid CAS:56-12-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0067100MG
Encompass_Preferred
Medchemexpress, HY-N0067 100mg γ-Aminobutyric acid CAS:56-12-2 γ-Aminobutyric acid (4-Aminobutyric acid) is a major inhibitory neurotransmitter in the adult mammalian brain [1] [2] , binding to the ionotropic GABA receptors ( GABA A receptors ) and metabotropic receptors ( GABA B receptors ) [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3378