Learn More
Foxx Life Sciences CPC AseptiQuik G Connector, 1/4" Hose Barb Genderless, Polycarbonate (PC), Class VI, AQG17004, 5/PK
Supplier:  Foxx Life Sciences AQG170045
Encompass_Preferred
Aseptiquik G Connectors provide quick and easy sterile connections, even in non-sterile environments—a critical capability for biopharmaceutical manufacturers. Featuring an easy to use three-step design and a wide range of options—including 1/8- to 1 1/2-inch size and genderless connections—the AseptiQuik family allows you to transfer media easily with less risk of error. Their robust, reliable performance eliminates the need for clamps, fixtures, or tube welders, giving you sterile, high-quality single-use connections every time.
Catalog No. 50-206-0521