Learn More
Foxx Life Sciences CPC AseptiQuik G HT Sanitary Connector, 1-1/2" Hose Barb Genderless, Polycarbonate (PC), Class VI, AQG33024HT, 25/PK
Catalog No. 502060541
Encompass_Preferred
AseptiquikG Connectors provide quick and easy sterile connections, even in non-sterile environments—a critical capability for biopharmaceutical manufacturers. Featuring an easy to use three-step design and a wide range of options—including 1/8- to 1 1/2-inch size and genderless connections—the AseptiQuik family allows you to transfer media easily with less risk of error. Their robust, reliable performance eliminates the need for clamps, fixtures, or tube welders, giving you sterile, high-quality single-use connections every time.
Catalog No. 50-206-0541 Supplier Foxx Life Sciences Supplier No. AQG33024HT25
AseptiquikG Connectors provide quick and easy sterile connections, even in non-sterile environments—a critical capability for biopharmaceutical manufacturers. Featuring an easy to use three-step design and a wide range of options—including 1/8- to 1 1/2-inch size and genderless connections—the AseptiQuik family allows you to transfer media easily with less risk of error. Their robust, reliable performance eliminates the need for clamps, fixtures, or tube welders, giving you sterile, high-quality single-use connections every time.