Learn More
Medchemexpress LLC HY-P7068 50ug Medchemexpress, G-CSF (CHO-expressed), Mouse CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP706850UG
Encompass_Preferred
Medchemexpress, HY-P7068 50ug G-CSF (CHO-expressed), Mouse CAS: Recombinant Mouse Granulocyte Colony-stimulating Factor (CHO-expressed) is a polypeptide growth factor that regulates the production of neutrophilic granulocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4311