Learn More
Sigma Aldrich Fine Chemicals Biosciences G CSf human Animal compone
Catalog No. 502906523
Encompass
Granulocyte colony stimulating factor (G-CSf) or colony stimulating factor 3 (CSf-3) is produced in response to specific stimulation by a variety of cells including macrophages fibroblasts endothelial cells and bone marrow stroma. Human and murine G-CSf are cross-species reactive. The gene encoding G-CSf is localized on human chromosome 17. Recombinant human G-CSf is an 18.7kDa protein consisting of 174 amino acid residues.
Catalog No. 50-290-6523 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP326310UG
Granulocyte colony stimulating factor (G-CSf) or colony stimulating factor 3 (CSf-3) is produced in response to specific stimulation by a variety of cells including macrophages fibroblasts endothelial cells and bone marrow stroma. Human and murine G-CSf are cross-species reactive. The gene encoding G-CSf is localized on human chromosome 17. Recombinant human G-CSf is an 18.7kDa protein consisting of 174 amino acid residues.