Learn More
Medchemexpress LLC G-CSF Human HEK293 100ug
SDP
Catalog No. 5000180382
Encompass_Preferred
G-CSF a pivotal cytokine regulates hematopoiesis by controlling the production differentiation and function of granulocytes and monocytes-macrophages This monomeric protein specifically promotes granulocyte development playing a crucial role in immune system regulation and blood cell formation G-CSF Protein Human (HEK293 Fc) is the recombinant human-derived G-CSF protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-80382 Supplier Medchemexpress LLC Supplier No. HYP73073100UG
G-CSF a pivotal cytokine regulates hematopoiesis by controlling the production differentiation and function of granulocytes and monocytes-macrophages This monomeric protein specifically promotes granulocyte development playing a crucial role in immune system regulation and blood cell formation G-CSF Protein Human (HEK293 Fc) is the recombinant human-derived G-CSF protein expressed by HEK293 with N-hFc labeled tag