Learn More
STEMCELL Technologies G-CSF Mobilized Human Peripheral Blood Mononuclear Cells, Frozen, Size: 5 x 10^6 cells
SDP
Catalog No. 501975816
Encompass
Start your assays with ready-to-use, ethically sourced, mobilized primary mononuclear cells. We help you get the cells you need with personalized service, custom products, flexible delivery times, and the option to reserve entire lots to prescreen cells for applications. Donors are mobilized with G-CSF, plerixafor, or G-CSF and plerixafor prior to collection. Isolated from leukapheresis samples using density gradient separation or red blood cell lysis and cryopreserved in CryoStorCS10 medium, cells are obtained using Institutional Review Board (IRB)-approved consent forms and protocols. Additional documentation and high-resolution HLA typing (Class I and Class II alleles and CMV status) are available upon request. ACDA is added as an anticoagulant. Contact techsupport@stemcell.com for donor specifications (BMI category, smoking status, ethnicity, etc.). Donors are screened for HIV-1, HIV-2, hepatitis B, and hepatitis C. Certain products are only available in select territories.
Catalog No. 50-197-5816 Supplier STEMCELL Technologies Supplier No. 70049.4
Add to Cart
Add to Cart
Start your assays with ready-to-use, ethically sourced, mobilized primary mononuclear cells. We help you get the cells you need with personalized service, custom products, flexible delivery times, and the option to reserve entire lots to prescreen cells for applications. Donors are mobilized with G-CSF, plerixafor, or G-CSF and plerixafor prior to collection. Isolated from leukapheresis samples using density gradient separation or red blood cell lysis and cryopreserved in CryoStorCS10 medium, cells are obtained using Institutional Review Board (IRB)-approved consent forms and protocols. Additional documentation and high-resolution HLA typing (Class I and Class II alleles and CMV status) are available upon request. ACDA is added as an anticoagulant. Contact techsupport@stemcell.com for donor specifications (BMI category, smoking status, ethnicity, etc.). Donors are screened for HIV-1, HIV-2, hepatitis B, and hepatitis C. Certain products are only available in select territories.