Learn More
STEMCELL Technologies G-CSF Mobilized Human Peripheral Blood CD34+ Cells, Frozen, Size: 1 x 10^7 cells
SDP
Supplier:  STEMCELL Technologies 70060.3
Encompass
Streamline your assays with ready-to-use, ethically sourced, mobilized primary CD34+ cells. We help you get the cells you need with personalized service, custom products, flexible delivery times, and the option to reserve entire lots to prescreen cells for applications. Donors are mobilized with G-CSF, plerixafor, or G-CSF and plerixafor prior to collection. Isolated from leukapheresis samples using positive immunomagnetic separation techniques and cryopreserved in serum-free medium, cells are obtained using Institutional Review Board (IRB)-approved consent forms and protocols. Additional documentation and high-resolution HLA typing (Class I and Class II alleles and CMV status) are available upon request. ACDA is added as an anticoagulant. Contact techsupport@stemcell.com for donor specifications (BMI category, smoking status, ethnicity, etc.). Donors are screened for HIV-1, HIV-2, hepatitis B, and hepatitis C. Certain products are only available in select territories.
Catalog No. 50-197-5826
Add to Cart
Request a Quote