Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0316310
Encompass
Granulocyte Colony-Stimulating Factor (G-CSF) also known as CSF-3 and MGI-1G is a cytokine and hormone belonging to the IL-6 superfamily. It is expressed by monocytes macrophages endothelial cells fibroblasts and bone marrow stroma. G-CSF stimulates the bone marrow to produce granulocytes and stem cells and specifically stimulates the proliferation and differentiation of the neutrophilic granulocyte lineage. G-CSF has been used to stimulate white blood cell production after chemotherapy. It has also been used to boost the number of hematopoietic stem cells after bone marrow transplantation.
Catalog No. 50-252-9761