Learn More
Medchemexpress LLC G-CSF Mouse HEK293 10ug
SDP
Catalog No. 5000184478
Encompass_Preferred
Recombinant mouse granulocyte colony-stimulating factor (G-CSF) produced in HEK293 cells and supplied as a lyophilized His-tagged protein for in vitro research. It promotes granulocyte proliferation and differentiation and is suitable for functional and cell-based assays.
  • Glycosylated protein expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • Lyophilized formulation for improved stability and shipping.
  • Low endotoxin level (<1 EU/μg) for cell-based assays.
  • High specific activity; ED50 ≤ 103.9 pg/mL in NFS-60 assay.
  • Store at -20°C; stable short-term at 4°C after reconstitution.
Catalog No. 50-001-84478 Supplier Medchemexpress LLC Supplier No. HYP7055410UG
Recombinant mouse granulocyte colony-stimulating factor (G-CSF) produced in HEK293 cells and supplied as a lyophilized His-tagged protein for in vitro research. It promotes granulocyte proliferation and differentiation and is suitable for functional and cell-based assays.
  • Glycosylated protein expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • Lyophilized formulation for improved stability and shipping.
  • Low endotoxin level (<1 EU/μg) for cell-based assays.
  • High specific activity; ED50 ≤ 103.9 pg/mL in NFS-60 assay.
  • Store at -20°C; stable short-term at 4°C after reconstitution.