Learn More
ACROBiosystems ACROBiosystems Biotinylated Human G-CSF R / CD114 Protein, Avitag™,His Tag
Catalog No. 501957445
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human G-CSF R, Avitag,His Tag (GCR-H82E4) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 621 (Accession # NP_000751).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 69.2 kDa. The protein migrates as 72-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human G-CSF, Tag Free (Cat. No. GCF-H5214) at 2 μg/mL (100 μL/well) can bind Biotinylated Human G-CSF R, Avitag,His Tag (Cat. No. GCR-H82E4) with a linear range of 0.156-1.25 μg/mL (QC tested).
Catalog No. 50-195-7445 Supplier ACROBiosystems Supplier No. GCRH82E4200UG
Recombinant Protein;200UG;Biotinylated Human G-CSF R, Avitag,His Tag (GCR-H82E4) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 621 (Accession # NP_000751).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 69.2 kDa. The protein migrates as 72-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human G-CSF, Tag Free (Cat. No. GCF-H5214) at 2 μg/mL (100 μL/well) can bind Biotinylated Human G-CSF R, Avitag,His Tag (Cat. No. GCR-H82E4) with a linear range of 0.156-1.25 μg/mL (QC tested).