Learn More
CPC Scientific H-Gly-DArg-Gly-Asp-Ser-Pro-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific FIBN010A
Encompass
ONE-LETTER SEQUENCE: GrGDSPMOLECULAR FORMULA: C22H37N9O10MOLECULAR WEIGHT:587.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Inflammation
Catalog No. 50-210-9286