Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences G-Fusion Cloning Kit
SCI_ED

Catalog No. S000743
Change view
Click to view available options
No. of Reactions:
10 Reactions
50 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
S000743 10 Reactions
S000754 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000743 Supplier G-Biosciences Supplier No. 7862199
Add to Cart
Add to Cart

G-Fusion is a simple, rapid and highly efficient directional cloning of one or more fragments of DNA into any vector.

The linearized vector can be generated by PCR or restriction enzymes. The PCR fragments can be generated with high fidelity DNA polymerase with primers having 15 to 18 bases of homology at their linear ends to where the product needs to fuse.

Specifications

Format Kit
No. of Reactions 10 Reactions
Product Type G-Fusion Cloning Kit
Quantity 20 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.