Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-Biosciences G-Fusion Cloning Kit

Description
The linearized vector can be generated by PCR or restriction enzymes. The PCR fragments can be generated with high fidelity DNA polymerase with primers having 15 to 18 bases of homology at their linear ends to where the product needs to fuse.
Specifications
Specifications
| Format | Kit |
| No. of Reactions | 50 Reactions |
| Product Type | G-Fusion Cloning Kit |
| Quantity | 100 μL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?