Learn More
Cayman Chemical G-Lactone 1g
SDP
Catalog No. 502992359
Encompass_Preferred
A bicyclic γ-lactone; a prostaglandin chiral synthon formed from an asymmetric Baeyer-Villiger oxidation reaction; has been used as a building block in the synthesis of HIV-1 protease inhibitors
Catalog No. 50-299-2359 Supplier Cayman Chemical Supplier No. 304881 G
A bicyclic γ-lactone; a prostaglandin chiral synthon formed from an asymmetric Baeyer-Villiger oxidation reaction; has been used as a building block in the synthesis of HIV-1 protease inhibitors