Learn More
Medchemexpress LLC (-)-G-Lactone | 43119-28-4 | ≥98.0% | 124.14 | 1 G
SDP
Catalog No. 5000403592
Encompass_Preferred
(-)-G-Lactone is a synthetic intermediate useful for prostaglandins synthesis. It is a drug intermediate and is for research use only. The appearance is a solid with an off-white to light yellow color.
  • Useful for prostaglandins synthesis
  • Drug intermediate
  • For research use only
  • Solid appearance
  • Off-white to light yellow color
  • Store at room temperature for 3 years
  • In solvent: -80°C for 2 years, -20°C for 1 year
Catalog No. 50-004-03592 Supplier Medchemexpress LLC Supplier No. HY430011G
(-)-G-Lactone is a synthetic intermediate useful for prostaglandins synthesis. It is a drug intermediate and is for research use only. The appearance is a solid with an off-white to light yellow color.
  • Useful for prostaglandins synthesis
  • Drug intermediate
  • For research use only
  • Solid appearance
  • Off-white to light yellow color
  • Store at room temperature for 3 years
  • In solvent: -80°C for 2 years, -20°C for 1 year