Learn More
Medchemexpress LLC (-)-G-Lactone | 43119-28-4 | ≥98.0% | 124.14 | 100 MG
SDP
Catalog No. 5000405124
Encompass_Preferred
(-)-G-Lactone is a synthetic intermediate useful for prostaglandins synthesis. It is intended for research use only.
  • Synthetic intermediate
  • Useful for prostaglandins synthesis
Catalog No. 50-004-05124 Supplier Medchemexpress LLC Supplier No. HY43001100MG
(-)-G-Lactone is a synthetic intermediate useful for prostaglandins synthesis. It is intended for research use only.
  • Synthetic intermediate
  • Useful for prostaglandins synthesis