Learn More
Medchemexpress LLC HY-B2194 1g Medchemexpress, γ-Oryzanol CAS:11042-64-1 Purity:>98%
SDP
Catalog No. 501933105
Encompass_Preferred
Medchemexpress, HY-B2194 1g γ-Oryzanol CAS:11042-64-1 γ-Oryzanol is a potent DNA methyltransferases ( DNMTs ) inhibitor in the striatum of mice. γ-Oryzanol significantly inhibits the activities of DNMT1 ( IC 50 =3.2 μM), DNMT3a ( IC 50 =22.3 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3105 Supplier Medchemexpress LLC Supplier No. HYB21941G
Medchemexpress, HY-B2194 1g γ-Oryzanol CAS:11042-64-1 γ-Oryzanol is a potent DNA methyltransferases ( DNMTs ) inhibitor in the striatum of mice. γ-Oryzanol significantly inhibits the activities of DNMT1 ( IC 50 =3.2 μM), DNMT3a ( IC 50 =22.3 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.