Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC G/P CLEANER

Catalog No. 502818359
Encompass

This concentrated cleaner is a high-performance, all-purpose solution for floors, walls, and other nonporous surfaces, functioning effectively as a hard surface cleaner. It is designed for industrial/occupational use and meets Green Seal™ Standard GS-37 based on effective performance, concentrated volume, minimized/recycled packaging, and protective limits on VOCs and human & environmental toxicity.

  • Liquid concentrate
  • Citrus scent
  • Red color
  • PH value 10-11
  • Density 8.65 lb/gal
  • Complete solubility in water
  • VOC less H2O and exempt solvents < 7 g/l
  • Packaged in 0.5 gal bottles, 4 per case

Catalog No. 50-281-8359 Supplier MSC Supplier No. 14162176
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This concentrated cleaner is a high-performance, all-purpose solution for floors, walls, and other nonporous surfaces, functioning effectively as a hard surface cleaner. It is designed for industrial/occupational use and meets Green Seal™ Standard GS-37 based on effective performance, concentrated volume, minimized/recycled packaging, and protective limits on VOCs and human & environmental toxicity.

  • Liquid concentrate
  • Citrus scent
  • Red color
  • PH value 10-11
  • Density 8.65 lb/gal
  • Complete solubility in water
  • VOC less H2O and exempt solvents < 7 g/l
  • Packaged in 0.5 gal bottles, 4 per case

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.