Learn More
CPC Scientific H-Gly-Arg-Gly-Asp-DSer-Pro-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific FIBN012A
Encompass
Inhibitor of cell attachment to fibronection but not vitronectin. ONE-LETTER SEQUENCE: GRGDsPMOLECULAR FORMULA: C22H37N9O10MOLECULAR WEIGHT:587.58STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [111844-24-7], RESEARCH AREA: Inflammation
Catalog No. 50-210-9290