Learn More
CPC Scientific H-Gly-Arg-Gly-Asp-Ser-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific FIBN013B
Encompass
Inhibitor of thrombin-induced platelet aggregation, derived from the cell binding region of fibronectin. This peptide competitively inhibits direct binding of fibroblasts to fibronectin.SEQUENCE: H-Gly-Arg-Gly-Asp-Ser-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: GRGDSMOLECULAR FORMULA: C17H30N8O9MOLECULAR WEIGHT: 490.47STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [96426-21-0]SYNONYMS: RESEARCH AREA: HematologyREFERENCES:M.J. Humphries et al., Science, 233, 467 (1986)SKU(s): FIBN-013A, FIBN-013B
Catalog No. 50-210-9293