Learn More
Wilson Wolf Manufacturing Corp G-Rex 6 Well Plate, 40 ml fill volume, 1 each
SDP
Catalog No. NC1000862
Encompass
Each well can expand 5 million cells into between 200 and 400 million cells in about 10 days with only one exchange of medium. Individual Well Specifications: 10cm2 gas permeable membrane surface area with 40 ml fill volume.
Catalog No. NC1000862 Supplier Wilson Wolf Manufacturing Corp Supplier No. 80240M
Each well can expand 5 million cells into between 200 and 400 million cells in about 10 days with only one exchange of medium. Individual Well Specifications: 10cm2 gas permeable membrane surface area with 40 ml fill volume.