Learn More
Wilson Wolf Manufacturing Corp G-Rex 6M Well Plate, 100ml
SDP
Catalog No. NC1498767
Encompass
Individual well specifications of 10 cm² gas permeable membrane. Expand 5 million cells into between 200 to 400 million cells in about 10 days with no medium exchange.
Catalog No. NC1498767 Supplier Wilson Wolf Manufacturing Corp Supplier No. 80660M
Individual well specifications of 10 cm² gas permeable membrane. Expand 5 million cells into between 200 to 400 million cells in about 10 days with no medium exchange.