Learn More
Wilson Wolf Manufacturing Corp G-REX10 GAS PERMEABLE CULT DEV
SDP
Catalog No. NC0518349
Encompass
Gas permeable culture device with a 10 cm2 surface and a 40 ml medium capacity. Typically generates 100 to 300 million cells after 2 medium exchanges. 2.0 inch diameter x 2.5 inch high.
Catalog No. NC0518349 Supplier Wilson Wolf Manufacturing Corp Supplier No. 80040S
Gas permeable culture device with a 10 cm2 surface and a 40 ml medium capacity. Typically generates 100 to 300 million cells after 2 medium exchanges. 2.0 inch diameter x 2.5 inch high.