Learn More
Wilson Wolf Manufacturing Corp G-REX100M-CS 3/CS
SDP
Supplier:  Wilson Wolf Manufacturing Corp 81100CS
Encompass
G-Rex®100M-CS (closed system), Sterile Fluid Path – P/N 81100-CS100cm² gas permeable membrane surface area with 1000 mL fill volume. Closed system with semi-automated liquid handling. Expand 50 million cells into between 2 to 4 billion cells in about 10 days with NO medium exchange. [Validated Sterile Fluid Path]
Catalog No. NC2006595