Learn More
Wilson Wolf Manufacturing Corp G-Rex10M Open System, Research Use Only, 3-pack Box
SDP
Catalog No. NC1520165
Encompass
This item has a minimum order quantity of 3 packs per supplier requirements. 10cm² gas permeable membrane surface area with 100 mL media capacity. Expand 5 million cells into between 200 to 400 million cells in about 10 days with NO medium exchange. [GAMMA IRRADIATED. For non-clinical, non-therapeutic, research use only.]
Catalog No. NC1520165 Supplier Wilson Wolf Manufacturing Corp Supplier No. 80110S
This item has a minimum order quantity of 3 packs per supplier requirements. 10cm² gas permeable membrane surface area with 100 mL media capacity. Expand 5 million cells into between 200 to 400 million cells in about 10 days with NO medium exchange. [GAMMA IRRADIATED. For non-clinical, non-therapeutic, research use only.]