Learn More
MESA LABS BOZEMAN MFG FACILITY SPORE STRIP STEAM LOG 6 100/pk
SDP
Supplier:  MESA LABS BOZEMAN MFG FACILITY 770136100/DEL
Encompass
G. stearothermophilus. Used for sterility testing in various sterilization environments, for both healthcare and industrial applications. Autoclave validations, EO Sterilization, Used in steam applications, Irradiation sterilization.
Catalog No. NC9749957