Learn More
Echelon Biosciences Research Labs G - Subtide - 5 mg,
SDP
Supplier:  Echelon Biosciences Research Labs 871155MG
Encompass_Preferred
G - Subtide - 5mg , Gln-Lys-Arg-Pro-Arg-Arg-Lys-Asp-Thr-Pro-OH Category/Application: Enzyme Substrates This item is non returnable
Catalog No. 50-190-9002