Learn More
Covaris Inc g-TUBE for DNA Shearing, 10/pk
SDP
Catalog No. NC0380758
Encompass
10 individually-wrapped g-TUBES for shearing DNa 6kb to 20 kb and one g-TUBE Prep Statio
Catalog No. NC0380758 Supplier Covaris Inc Supplier No. 520079
10 individually-wrapped g-TUBES for shearing DNa 6kb to 20 kb and one g-TUBE Prep Statio