Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC G0-C14 | 1510653-27-6 | C106H216N10O10 | 100 MG
SDP

Catalog No. 5000372630
Encompass_Preferred

G0-C14 is a cationic lipid-like compound, an alkyl-modified polyamidoamine (PAMAM) dendrimer, utilized in the preparation of macrophage-targeted nanoparticles (NPs) for agent and vaccine delivery. It demonstrates high entrapment efficiency (above 95%) of mRNA and pDNA. It is intended for research use only.

  • Cationic lipid-like compound
  • Alkyl-modified polyamidoamine (PAMAM) dendrimer
  • Used in preparation of macrophage-targeted nanoparticles for agent and vaccine delivery
  • High entrapment efficiency (above 95%) of mRNA and pDNA
  • Purity of 98.0%
  • Appears as a colorless to light yellow oil
  • Soluble in DMSO at 100 mg/mL
  • Requires ultrasonic and warming/heating for dissolution in DMSO due to hygroscopic nature
  • Can be formulated with various in vivo dissolution protocols

Catalog No. 50-003-72630 Supplier Medchemexpress LLC Supplier No. HY152229100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

G0-C14 is a cationic lipid-like compound, an alkyl-modified polyamidoamine (PAMAM) dendrimer, utilized in the preparation of macrophage-targeted nanoparticles (NPs) for agent and vaccine delivery. It demonstrates high entrapment efficiency (above 95%) of mRNA and pDNA. It is intended for research use only.

  • Cationic lipid-like compound
  • Alkyl-modified polyamidoamine (PAMAM) dendrimer
  • Used in preparation of macrophage-targeted nanoparticles for agent and vaccine delivery
  • High entrapment efficiency (above 95%) of mRNA and pDNA
  • Purity of 98.0%
  • Appears as a colorless to light yellow oil
  • Soluble in DMSO at 100 mg/mL
  • Requires ultrasonic and warming/heating for dissolution in DMSO due to hygroscopic nature
  • Can be formulated with various in vivo dissolution protocols

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.