Learn More
Medchemexpress LLC G0-C14 | 1510653-27-6 | C106H216N10O10 | 25 MG
SDP
Catalog No. 5000374591
Encompass_Preferred
G0-C14 is a cationic lipid-like compound, an alkyl-modified polyamidoamine (PAMAM) dendrimer. It is utilized in the preparation of macrophage-targeted nanoparticles (NPs) for agent and vaccine delivery.
  • Strong entrapment of mRNA and pDNA with over 95% encapsulation efficiency.
  • Appears as an oil.
  • Colorless to light yellow in appearance.
  • Purity of 98.0%.
Catalog No. 50-003-74591 Supplier Medchemexpress LLC Supplier No. 25MGHY152229
G0-C14 is a cationic lipid-like compound, an alkyl-modified polyamidoamine (PAMAM) dendrimer. It is utilized in the preparation of macrophage-targeted nanoparticles (NPs) for agent and vaccine delivery.
  • Strong entrapment of mRNA and pDNA with over 95% encapsulation efficiency.
  • Appears as an oil.
  • Colorless to light yellow in appearance.
  • Purity of 98.0%.