Learn More
Apexbio Technology LLC G007-LK 1380672-07-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062765
Encompass_Preferred
G007-LK (CAS 1380672-07-0) is a small molecule inhibitor that selectively targets tankyrase 1 and 2 (TNKS1/2) enzymes within the poly(ADP-ribose) polymerase family that modulate protein complexes through ADP-ribosylation G007-LK inhibits TNKS1 and TNKS2 with IC50 values of 46 nM and 25 nM respectively reducing their auto-poly(ADP-ribosylation) activity Treatment with G007-LK decreases -catenin levels in APC-mutated colorectal cancer cells by promoting formation of -catenin degradation complexes In xenograft mouse models using COLO-320DM cells G007-LK administration suppresses tumor growth dose-dependently indicating its utility in oncology research focusing on the Wnt/ -catenin pathway
Catalog No. 50-000-62765 Supplier Apexbio Technology LLC Supplier No. B58305.1
G007-LK (CAS 1380672-07-0) is a small molecule inhibitor that selectively targets tankyrase 1 and 2 (TNKS1/2) enzymes within the poly(ADP-ribose) polymerase family that modulate protein complexes through ADP-ribosylation G007-LK inhibits TNKS1 and TNKS2 with IC50 values of 46 nM and 25 nM respectively reducing their auto-poly(ADP-ribosylation) activity Treatment with G007-LK decreases -catenin levels in APC-mutated colorectal cancer cells by promoting formation of -catenin degradation complexes In xenograft mouse models using COLO-320DM cells G007-LK administration suppresses tumor growth dose-dependently indicating its utility in oncology research focusing on the Wnt/ -catenin pathway