Learn More
Selleck Chemical LLC G007-LK 5mg 1380672-07-0
SDP
Supplier:  Selleck Chemical LLC S72395MG
Encompass_Preferred
G007-LK is a potent and selective tankyrase inhibitor with IC50 of 46 nM and 25 nM for TNKS1/2, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1107580