Learn More
Grainger G0361 Pgbrd Panel 3/4 x32 x48 in Crcl Slttd
Catalog No. 502751418
Encompass_Preferred
The Grainger 14K265 is a Westward brand 1/4-inch drive socket set. It contains both standard and deep sockets ranging in size from 4mm to 14mm. The sockets have a chrome vanadium steel construction with a black phosphate finish. The set comes packaged in a molded plastic case for storage and portability. This 73 piece socket set provides a wide range of metric sockets for automotive and machinery applications.
Catalog No. 50-275-1418 Supplier Grainger Supplier No. 14K265
The Grainger 14K265 is a Westward brand 1/4-inch drive socket set. It contains both standard and deep sockets ranging in size from 4mm to 14mm. The sockets have a chrome vanadium steel construction with a black phosphate finish. The set comes packaged in a molded plastic case for storage and portability. This 73 piece socket set provides a wide range of metric sockets for automotive and machinery applications.