Learn More
Grainger G0362 Pgbrd Panel 3/4 x32 x96 in Crcl Slttd
Catalog No. 502750363
Encompass_Preferred
The FisherbrandTM 502750236 is a polypropylene microcentrifuge tube rack that holds 96 tubes of 1.5mL capacity. It has alphanumeric grid references for easy sample identification and location. The rack is made of durable autoclavable polypropylene and can withstand temperatures from -20°C to 121°C. It has dimensions of 127mm x 127mm x 45mm. This microcentrifuge tube rack allows for organized storage and easy access to samples in 1.5mL tubes. Its sturdy construction and alphanumeric grid make it useful for applications requiring sample tracking and identification.
Catalog No. 50-275-0363 Supplier Grainger Supplier No. 14K274
The FisherbrandTM 502750236 is a polypropylene microcentrifuge tube rack that holds 96 tubes of 1.5mL capacity. It has alphanumeric grid references for easy sample identification and location. The rack is made of durable autoclavable polypropylene and can withstand temperatures from -20°C to 121°C. It has dimensions of 127mm x 127mm x 45mm. This microcentrifuge tube rack allows for organized storage and easy access to samples in 1.5mL tubes. Its sturdy construction and alphanumeric grid make it useful for applications requiring sample tracking and identification.