Learn More
Cayman Chemical G-1, 881639-98-1, 10 mg
SDP
Supplier:  Cayman Chemical 1000893310
Encompass_Preferred
Molecular Formula C21H18BrNO3, Purity ≥98%, Formula Weight 412.3; G-1 is a nonsteroidal, high-affinity, selective agonist of GPR30 that binds with a Ki value of 11 nM. Competitive binding studies in endoplasmic reticulum - (ER-) and ER-expressing cells yielded Ki values for estradiol of 0.30 and 0.38 nM, respectively, with no substantial binding of G-1 at 1 M.{14210} The discovery of G-1, a compound that does not bind classical ERs, should facilitate further physiological experiments to define the role of GPR30 in vivo.
Catalog No. NC0591839