Learn More
Medchemexpress LLC G1-OC2-K3-E10 | 2933216-12-5 | 98.9% | 942.49 | 1 MG
SDP
Catalog No. 5000391696
Encompass_Preferred
G1-OC2-K3-E10 is an ionizable lipid designed for the delivery of mRNA in lipid nanoparticles (LNPs). This product is intended for research use only.
  • Used for delivery of mRNA in lipid nanoparticles (LNPs)
  • For research use only
  • Not intended for human use or sale to patients
  • Store at -20°C, protected from light and under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen
Catalog No. 50-003-91696 Supplier Medchemexpress LLC Supplier No. HY1605801MG
G1-OC2-K3-E10 is an ionizable lipid designed for the delivery of mRNA in lipid nanoparticles (LNPs). This product is intended for research use only.
  • Used for delivery of mRNA in lipid nanoparticles (LNPs)
  • For research use only
  • Not intended for human use or sale to patients
  • Store at -20°C, protected from light and under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen