Learn More
Abcam G10, STING signaling activator, 5MG
Catalog No. 502672646
Encompass_Preferred
MW 430.9 Da, Purity >98%. G10 is an indirect activator or stimulator of interferon genes (STING) signaling. It induces transcription of genes dependent on IRF3 and IFN, which are activated by STING, but has no effect on non-STING activated NF-κB-dependent transcription of IL-8, IL-1β, or MIP-1α in human fibroblasts. However, G10 does increase expression of NF-κB-dependent genes in human peripheral blood mononuclear cells (PBMCs) and human umbilical microvascular endothelial cells (HUMECs). In vitro, it prevents replication of the chikungunya and Venezuelan equine encephalitis alphaviruses (IC₉₀s = 8.01 and 24.57 μM, respectively). G10 (100 μM) increases phosphorylation of IRF3, which is lost in cells lacking STING.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2646 Supplier Abcam Supplier No. AB2303755MG
MW 430.9 Da, Purity >98%. G10 is an indirect activator or stimulator of interferon genes (STING) signaling. It induces transcription of genes dependent on IRF3 and IFN, which are activated by STING, but has no effect on non-STING activated NF-κB-dependent transcription of IL-8, IL-1β, or MIP-1α in human fibroblasts. However, G10 does increase expression of NF-κB-dependent genes in human peripheral blood mononuclear cells (PBMCs) and human umbilical microvascular endothelial cells (HUMECs). In vitro, it prevents replication of the chikungunya and Venezuelan equine encephalitis alphaviruses (IC₉₀s = 8.01 and 24.57 μM, respectively). G10 (100 μM) increases phosphorylation of IRF3, which is lost in cells lacking STING.
The product is subject to the following: Abcam Restricted Use Statement