Learn More
Perkin Elmer US LLC G10 Test Sample - Qty. 5
Catalog No. 502105546
Encompass_Preferred
The Measuring System requires the following calibration test samples to ensure optimal performance: PMMA, Steel, PVC, HDPE, Polycarb, PET Film, and G10 Epoxy. The test samples come standard with this DMA 8000 system, but they can also be ordered individually, as needed.
Catalog No. 50-210-5546 Supplier Perkin Elmer US LLC Supplier No. N5330344
The Measuring System requires the following calibration test samples to ensure optimal performance: PMMA, Steel, PVC, HDPE, Polycarb, PET Film, and G10 Epoxy. The test samples come standard with this DMA 8000 system, but they can also be ordered individually, as needed.