Learn More
Electron Microscopy Sciences Counting Pattern Graticle G14, 19mm, Counting pattern
SDP
Catalog No. 5025127
Encompass_Preferred
Simple counting for geological and soil analysis. Reference: L.G. Briarty. "Stereology: Methods for Quantitative Light and Electron Microscopy." Sci. Prog. Oxf. 1975 62; 1-32.
Size: 19mm
Divisions: Counting pattern
Surface: Chrome image
Catalog No. 50-251-27 Supplier Electron Microscopy Sciences Supplier No. 6809219
Simple counting for geological and soil analysis. Reference: L.G. Briarty. "Stereology: Methods for Quantitative Light and Electron Microscopy." Sci. Prog. Oxf. 1975 62; 1-32.
Size: 19mm
Divisions: Counting pattern
Surface: Chrome image