Learn More
Electron Microscopy Sciences Counting Pattern Graticle G14, 21mm, Counting pattern
SDP
Catalog No. 5025126
Encompass_Preferred
Simple counting for geological and soil analysis. Reference: L.G. Briarty. "Stereology: Methods for Quantitative Light and Electron Microscopy." Sci. Prog. Oxf. 1975 62; 1-32.
Size: 21mm
Divisions: Counting pattern
Surface: Chrome image
Catalog No. 50-251-26 Supplier Electron Microscopy Sciences Supplier No. 6809221
Simple counting for geological and soil analysis. Reference: L.G. Briarty. "Stereology: Methods for Quantitative Light and Electron Microscopy." Sci. Prog. Oxf. 1975 62; 1-32.
Size: 21mm
Divisions: Counting pattern
Surface: Chrome image