Learn More
Medchemexpress LLC HY-133916 5mg , G140 CAS:2369751-07-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1339165MG
Encompass_Preferred
HY-133916 5mg G140 CAS: 2369751-07-3 G140 is a potent and selective inhibitor of cyclic GMP-AMP synthase (cGAS), with IC50s of 14.0 nM and 442 nM for h-cGAS and m-cGAS, respectively. G140 has anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9494