Learn More
Medchemexpress LLC HY-103449 5mg Medchemexpress, G15 CAS:1161002-05-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1034495MG
Encompass_Preferred
Medchemexpress, HY-103449 5mg G15 CAS:1161002-05-6 G15 is a high affinity and selective G-protein-coupled estrogen receptor (GPER/GPR30) antagonist with a K i of 20 nM [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2245