Learn More
Selleck Chemical LLC G15 GRB-G15-5mg
SDP
Catalog No. 502389945
Encompass_Preferred
G15 is a cell-permeable high affinity and selective G-protein coupled estrogen receptor 1 (GPER, GPR30) antagonist with an affinity of approximately 20 nM that displays no affinity for ERα and ERβ at concentrations up to 10 μM.
Catalog No. 50-238-9945 Supplier Selleck Chemical LLC Supplier No. S66515MG
G15 is a cell-permeable high affinity and selective G-protein coupled estrogen receptor 1 (GPER, GPR30) antagonist with an affinity of approximately 20 nM that displays no affinity for ERα and ERβ at concentrations up to 10 μM.