Learn More
Selleck Chemical LLC G150 S8944-25MG
SDP
Catalog No. 5000024202
Encompass_Preferred
G150 is a potent and highly selective human cyclic GMP-AMP synthase (cGAS) inhibitor with IC50 of 10 2 nM
Catalog No. 50-000-24202 Supplier Selleck Chemical LLC Supplier No. S894425MG
G150 is a potent and highly selective human cyclic GMP-AMP synthase (cGAS) inhibitor with IC50 of 10 2 nM