Learn More
Medchemexpress LLC G150 | 2369751-30-2 | 99.2% | 100 MG
SDP
Catalog No. 5000341962
Encompass_Preferred
G150 is a highly selective human cyclic GMP-AMP synthase (h-cGAS) inhibitor with an IC50 of 10.2 nM. It represses dsDNA-triggered interferon expression and can be used for research on inflammatory conditions.
  • Highly selective h-cGAS inhibitor
  • IC50 of 10.2 nM
  • Represses dsDNA-triggered interferon expression
  • Useful for research on inflammatory conditions
Catalog No. 50-003-41962 Supplier Medchemexpress LLC Supplier No. HY128583100MG
G150 is a highly selective human cyclic GMP-AMP synthase (h-cGAS) inhibitor with an IC50 of 10.2 nM. It represses dsDNA-triggered interferon expression and can be used for research on inflammatory conditions.
  • Highly selective h-cGAS inhibitor
  • IC50 of 10.2 nM
  • Represses dsDNA-triggered interferon expression
  • Useful for research on inflammatory conditions