Learn More
Selleck Chemical LLC G1T38
SDP
Catalog No. 5000002841
Encompass_Preferred
G1T38 (Lerociclib) is a novel potent selective and orally bioavailable CDK4/6 inhibitor with IC50 values of 0 001 M 0 002 M and 0 028 M for CDK4 CDK6 and CDK9 respectively
Catalog No. 50-000-02841 Supplier Selleck Chemical LLC Supplier No. S85682MG
G1T38 (Lerociclib) is a novel potent selective and orally bioavailable CDK4/6 inhibitor with IC50 values of 0 001 M 0 002 M and 0 028 M for CDK4 CDK6 and CDK9 respectively